Search
Search

B4galt7| Gene Symbol: B4galt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146045.1 | NP_666157.1 | 4 | 721 | ||
| GenBank mRNA | AK141739.1 | - | 4 | 685 | ||
| AK146855.1 | - | 4 | 768 | |||
| AK171600.1 | - | 4 | 737 | |||
| BC027195.1 | - | 4 | 721 | 4224150 | ||
| BC055703.1 | - | 4 | 936 | 6403624 | ||