Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165952.1 | NP_001159424.1 | 2 | 1598 | ||
NM_001165953.1 | NP_001159425.1 | 2 | 1598 | |||
NM_194343.2 | NP_919324.2 | 2 | 1598 | |||
GenBank mRNA | AK076600.1 | - | 2 | 1174 | ||
AK150574.1 | - | 2 | 1599 | |||
AK153571.1 | - | 2 | 1599 | |||
BC057358.1 | - | 2 | 1476 | 6835531 |