Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145210.2 | NP_660211.2 | 4 | 864 | ||
GenBank mRNA | AB067531.1 | - | 4 | 838 | ||
AK087823.1 | - | 4 | 864 | |||
AY055830.1 | - | 4 | 837 | |||
AY055831.1 | - | 4 | 837 | |||
BC053721.1 | - | 4 | 861 | 30053184 | ||
BC094918.1 | - | 4 | 854 | 30050168 | ||
BC108956.1 | - | 4 | 860 | 40046020 |