Search
Search

Atg4d| Gene Symbol: Atg4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153583.10 | NP_705811.8 | 10 | 1569 | ||
| GenBank mRNA | AK036282.1 | - | 10 | 1557 | ||
| AK036571.1 | - | 10 | 1558 | |||
| AK081002.1 | - | 10 | 1556 | |||
| AK081605.1 | - | 10 | 1547 | |||
| AK087258.1 | - | 10 | 1761 | |||
| AK154130.1 | - | 10 | 1562 | |||
| AK213738.1 | - | 1 | 15 | |||
| BC030861.1 | - | 10 | 1546 | 4208700 | ||
| BC069851.1 | - | 10 | 1521 | 6490007 | ||