Search
Search

Ing4| Gene Symbol: Ing4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133345.2 | NP_579923.1 | 1, 2 | 44 | ||
| GenBank mRNA | AK002821.1 | - | 1, 2 | 46 | ||
| AK009267.1 | - | 1, 2 | 45 | |||
| AK031633.1 | - | 1, 2 | 49 | |||
| AK046126.1 | - | 1, 2 | 57 | |||
| AK050522.1 | - | 1, 2 | 57 | |||
| AY035880.1 | - | 1, 2 | 31 | |||
| AY035881.1 | - | 1, 2 | 31 | |||
| AY036107.1 | - | 1, 2 | 31 | |||
| BC009127.1 | - | 1, 2 | 32 | 3708722 | ||