Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010836.3 | NP_001010836.1 | 3 | 240 | ||
GenBank mRNA | AK146098.1 | - | 3 | 216 | ||
AK170535.1 | - | 3 | 241 | |||
AY869711.1 | - | 3 | 210 |