Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198635.3 | NP_941037.2 | 5 | 471 | ||
GenBank mRNA | AK041995.1 | - | 2 | 215 | ||
AK133270.1 | - | 5 | 472 | |||
BC139037.1 | - | 2 | 76 | |||
BC139038.1 | - | 2 | 76 | |||
BC145517.1 | - | 2 | 76 | |||
BC145518.1 | - | 2 | 76 | |||
BC145519.1 | - | 2 | 76 |