Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nanos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194064.2 | NP_918953.2 | 1 | 471 | ||
GenBank mRNA | AB095972.1 | - | 1 | 471 | ||
JQ936992.1 | - | 1 | 401 |