Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024850.2 | NP_001020021.1 | 1 | 388 | ||
GenBank mRNA | AK146090.1 | - | 1 | 370 | ||
BC100366.1 | - | 1 | 332 | 30524902 | ||
BC132474.1 | - | 1 | 319 | 40130751 | ||
BC132500.1 | - | 1 | 319 | 40130777 | ||
DQ058647.1 | - | 1 | 388 |