Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181649.6 | NP_857632.4 | 4 | 494 | ||
GenBank mRNA | AK009084.1 | - | 4 | 381 | ||
AK148331.1 | - | 4 | 496 | |||
BC047998.1 | - | 4 | 408 | 6511097 | ||
BC092522.1 | - | 4 | 421 | 30648550 | ||
BC138310.1 | - | 4 | 389 | 8861330 | ||
BY741452.1 | - | 4 | 495 |