Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018777.4 | NP_061247.1 | 3 | 135 | ||
GenBank mRNA | AF087824.1 | - | 1 | 12 | ||
AF125305.1 | - | 1 | 12 | |||
AK010560.1 | - | 3 | 136 | |||
AK010682.1 | - | 3 | 130 | |||
BC005718.1 | - | 3 | 112 | 3594823 | ||
BC050138.1 | - | 3 | 118 | 30028814 | ||
DQ864649.1 | - | 1 | 12 |