Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020044.3 | NP_064428.1 | 9 | 519 | ||
NM_022964.4 | NP_075253.2 | 8 | 483 | |||
GenBank mRNA | AF257136.1 | - | 8 | 463 | ||
AK134721.1 | - | 6 | 211 | |||
AK138953.1 | - | 9 | 478 | |||
AK143533.1 | - | 8 | 484 | |||
AK144261.1 | - | 9 | 458 | |||
AK162321.1 | - | 8 | 442 | |||
AY190024.1 | - | 9 | 484 | |||
BC005804.1 | - | 8 | 412 | 3154073 |