Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020605.3 | NP_065630.1 | 1, 2 | 442 | ||
GenBank mRNA | AB024449.1 | - | 1, 2 | 438 | ||
AK045706.1 | - | 1, 2 | 429 | |||
AK046136.1 | - | 435 | ||||
AK082880.1 | - | 1, 2 | 431 | |||
BC103682.1 | - | 1, 2 | 397 | 40047806 | ||
BC104736.1 | - | 1, 2 | 397 | 40047803 | ||
BC105307.1 | - | 1, 2 | 397 | 40047801 | ||
BY718602.1 | - | 1, 2 | 453 |