Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025375.3 | NP_079651.2 | 5 | 462 | ||
GenBank mRNA | AF412035.1 | - | 4 | 234 | ||
AK002497.1 | - | 3 | 556 | |||
AK003386.1 | - | 4 | 235 | |||
AK008966.1 | - | 2 | 67 | |||
AK011005.1 | - | 4 | 242 | |||
AK151986.1 | - | 4 | 243 | |||
BC093484.1 | - | 4 | 212 | 6735279 | ||
BC117986.1 | - | 5 | 462 | 40102067 | ||
BC117987.1 | - | 5 | 462 | 40102068 |