Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025995.2 | NP_080271.2 | 4, 5 | 1135 | ||
GenBank mRNA | AK011820.1 | - | 4, 5 | 1133 | ||
AK135916.1 | - | 4, 5 | 1089 | |||
AK160201.1 | - | 4, 5 | 1091 | |||
BC053434.1 | - | 4, 5 | 1059 | 6336178 |