Search
Search

Rgs10| Gene Symbol: Rgs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026418.2 | NP_080694.1 | 5 | 579 | ||
| GenBank mRNA | AK009283.1 | - | 5 | 580 | ||
| AK009686.1 | - | 5 | 581 | |||
| AK017314.1 | - | 5 | 580 | |||
| AK021202.1 | - | 5 | 583 | |||
| AK088009.1 | - | 5 | 578 | |||
| BC109178.1 | - | 5 | 564 | 40049527 | ||
| BC109179.1 | - | 5 | 569 | |||
| EF410104.1 | - | 376 | ||||