Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026464.2 | NP_080740.2 | 5 | 627 | ||
GenBank mRNA | AK018469.1 | - | 5 | 608 | ||
AK075958.1 | - | 5 | 627 | |||
AK146093.1 | - | 5 | 646 | |||
BC011484.1 | - | 3 | 293 | |||
BC064750.1 | - | 5 | 586 | |||
BC131932.1 | - | 5 | 621 | 40130209 | ||
BC131934.1 | - | 5 | 621 | 40130211 |