Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027480.3 | NP_081756.4 | 17 | 1979 | ||
GenBank mRNA | AK019475.1 | - | 4 | 645 | ||
AK043638.1 | - | 15 | 3061 | |||
AK173316.1 | - | 15 | 2945 | |||
BC017633.1 | - | 4 | 650 | |||
BC048236.1 | - | 16 | 1884 | 6313999 |