Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cyth4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028195.3 | NP_082471.2 | 12 | 1155 | ||
GenBank mRNA | AF079972.1 | - | 12 | 1086 | ||
AK010908.1 | - | 2 | 88 | |||
AK152306.1 | - | 12 | 1061 | |||
AK152909.1 | - | 12 | 1061 | |||
AK157026.1 | - | 12 | 1061 | |||
AK220198.1 | - | 12 | 1084 | |||
BC018505.1 | - | 11 | 979 | |||
BC139054.1 | - | 12 | 1128 | 8862076 | ||
BC139056.1 | - | 12 | 1128 | 8862078 | ||
BC145528.1 | - | 12 | 1155 |