Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175271.4 | NP_780480.2 | 3 | 1159 | ||
GenBank mRNA | AK045289.1 | - | 3 | 1129 | ||
AK051709.1 | - | 3 | 1130 | |||
AY255540.1 | - | 1 | 139 | |||
BC019743.1 | - | 1 | 396 | |||
BC052178.1 | - | 3 | 1150 | |||
BC069178.1 | - | 3 | 1071 | |||
BC138893.1 | - | 3 | 960 | 8861915 | ||
BC138894.1 | - | 3 | 960 | 8861916 |