Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007523.2 | NP_031549.2 | 4 | 485 | ||
GenBank mRNA | AF402617.1 | - | 3 | 232 | ||
AK089220.1 | - | 4 | 423 | |||
AK200878.1 | - | 2 | 125 | |||
AK211584.1 | - | 2 | 80 | |||
BC057589.1 | - | 4 | 437 | 6411580 | ||
CX564692.1 | - | 4 | 418 | |||
Y13231.1 | - | 4 | 483 |