Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008967.3 | NP_032993.2 | 1 | 1071 | ||
GenBank mRNA | AK033567.1 | - | 2 | 463 | ||
AK154272.1 | - | 1 | 1037 | |||
BC094386.1 | - | 1 | 1045 | 30608457 | ||
D26157.1 | - | 1 | 1062 |
Set Membership: |
G-protein coupled receptor |