Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144853.3 | NP_659102.1 | 2 | 412 | ||
GenBank mRNA | AF442733.1 | - | 2 | 147 | ||
AK031840.1 | - | 2 | 436 | |||
AK158088.1 | - | 2 | 470 | |||
AK163137.1 | - | 457 | ||||
AK171789.1 | - | 949 | ||||
AK171936.1 | - | 2 | 437 | |||
AY061854.1 | - | 2 | 434 | |||
BC099957.1 | - | 2 | 412 | 6431603 | ||
BC125594.1 | - | 2 | 206 | 40130109 | ||
BC125596.1 | - | 2 | 206 | 40130111 |