Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Parl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005767.4 | NP_001005767.1 | 2 | 208 | ||
GenBank mRNA | AK039142.1 | - | 2 | 187 | ||
AK150980.1 | - | 2 | 216 | |||
AK151652.1 | - | 2 | 207 | |||
AK179606.1 | - | 2 | 73 | |||
AK181973.1 | - | 2 | 74 | |||
AK207637.1 | - | 2 | 217 | |||
AK216516.1 | - | 2 | 73 | |||
BC083153.1 | - | 2 | 208 | 30728006 |