Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025602.3 | NP_079878.1 | 4 | 616 | ||
GenBank mRNA | AK012917.1 | - | 4 | 599 | ||
AK019970.1 | - | 4 | 617 | |||
AK135550.1 | - | 4 | 641 | |||
BC027401.1 | - | 4 | 598 | 4948072 |