Search
Search

Commd4| Gene Symbol: Commd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025417.2 | NP_079693.1 | 4 | 187 | ||
| GenBank mRNA | AK004174.1 | - | 4 | 188 | ||
| AK009802.1 | - | 4 | 186 | |||
| AK172194.1 | - | 3 | 1299 | |||
| BC033438.1 | - | 3 | 173 | 3995599 | ||
| BC049222.1 | - | 4 | 186 | 2649359 | ||
| BC055798.1 | - | 3 | 1278 | |||