Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025901.3 | NP_080177.1 | 3 | 370 | ||
GenBank mRNA | AK005152.1 | - | 3 | 274 | ||
AK008637.1 | - | 3 | 242 | |||
AK010875.1 | - | 3 | 308 | |||
AK021353.1 | - | 3 | 305 | |||
AK031110.1 | - | 3 | 305 | |||
AK088206.1 | - | 3 | 270 | |||
AK161764.1 | - | 1 | 13 | |||
BC025898.1 | - | 3 | 386 |