Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190734.1 | NP_001177663.1 | 10 | 736 | ||
NM_178896.5 | NP_849227.1 | 9 | 801 | |||
GenBank mRNA | AK033559.1 | - | 9 | 746 | ||
AK076991.1 | - | 10 | 737 | |||
BC060213.1 | - | 9 | 743 | |||
BC108346.1 | - | 9 | 703 | 6310392 | ||
BC137627.1 | - | 9 | 745 | 8860646 | ||
BC137628.1 | - | 9 | 745 | 8860647 |