Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Imp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133976.2 | NP_598737.1 | 1 | 436 | ||
GenBank mRNA | AK004456.1 | - | 1 | 463 | ||
BC009145.1 | - | 1 | 439 | 2631069 |
Gene Symbol: 4933427G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030236.1 | - | 705 |