Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139142.2 | NP_631881.1 | 5 | 662 | ||
GenBank mRNA | AJ428067.1 | - | 675 | |||
AJ964961.1 | - | 5 | 636 | |||
AK043297.1 | - | 5 | 673 | |||
AK080979.1 | - | 673 | ||||
AK140445.1 | - | 673 | ||||
AK160468.1 | - | 673 | ||||
BC118933.2 | - | 5 | 662 | 40090800 |