Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014981.1 | NP_001014981.1 | 13 | 1873 | ||
GenBank mRNA | AF188123.1 | - | 12 | 1635 | ||
AF188124.1 | - | 12 | 1635 | |||
AK122304.1 | - | 5 | 762 | |||
AK154451.1 | - | 13 | 1853 | |||
BC026618.1 | - | 13 | 1792 | |||
BC125573.1 | - | 13 | 1741 | 40130088 | ||
BC137785.1 | - | 13 | 1741 | 8860804 |