Search
Search

Carns1
LOC102631780| Gene Symbol: Carns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134148.2 | NP_598909.2 | 5 | 796 | ||
| GenBank mRNA | AK129347.1 | - | 5 | 771 | ||
| Gene Symbol: LOC102631780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087383.1 | - | 1 | 2008 | ||