Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033223.2 | NP_001028395.1 | 4 | 361 | ||
NM_001039354.1 | NP_001034443.1 | 4 | 403 | |||
NM_001284329.1 | NP_001271258.1 | 4 | 1120 | |||
GenBank mRNA | AK077191.1 | - | 4 | 1120 | ||
AK160671.1 | - | 4 | 310 | |||
BC029721.1 | - | 4 | 309 | |||
BC105965.1 | - | 3 | 256 | |||
BG288381.1 | - | 4 | 341 | |||
BU923246.1 | - | 4 | 370 |