Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ibtk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081282.2 | NP_001074751.1 | 8 | 1679 | ||
GenBank mRNA | AK030895.1 | - | 8 | 1642 | ||
AK037129.1 | - | 8 | 1670 | |||
AK050176.1 | - | 8 | 1671 | |||
AK129353.1 | - | 5 | 557 | |||
AK136872.1 | - | 8 | 1660 | |||
BC060722.1 | - | 8 | 1650 | |||
BC137798.1 | - | 8 | 1568 | 8860817 |