Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssc4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160366.1 | NP_001153838.1 | 6 | 1081 | ||
GenBank mRNA | BC117747.1 | - | 3 | 588 | ||
BC126947.1 | - | 3 | 588 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC068293.1 | - | 303 | Chr. - - [-] on Build |