Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Braf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139294.5 | NP_647455.3 | 21 | 2467 | ||
GenBank mRNA | AF454556.1 | - | 9 | 1314 | ||
AF454557.1 | - | 9 | 1314 | |||
AF454558.1 | - | 9 | 1314 | |||
AF454559.1 | - | 9 | 1314 | |||
AK140431.1 | - | 21 | 2468 |