Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021600.3 | NP_067611.2 | 7 | 810 | ||
GenBank mRNA | AK081344.1 | - | 6 | 796 | ||
BC052153.1 | - | 7 | 755 | 6336578 | ||
BC141376.1 | - | 7 | 738 | 9055989 | ||
BC141387.1 | - | 7 | 738 | 9056000 | ||
K02582.1 | - | 7 | 721 | |||
L10076.1 | - | 7 | 770 |