Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054070.3 | NP_473411.2 | 6 | 619 | ||
GenBank mRNA | AF329695.1 | - | 6 | 560 | ||
AK159647.1 | - | 6 | 583 | |||
AK167964.1 | - | 6 | 594 | |||
AK168244.1 | - | 6 | 593 | |||
AK218541.1 | - | 2 | 89 | |||
BC056978.1 | - | 6 | 578 | 6821746 | ||
BC060064.1 | - | 6 | 578 |