Search
Search

Adk| Gene Symbol: Adk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243041.1 | NP_001229970.1 | 11 | 1136 | ||
| NM_134079.4 | NP_598840.1 | 11 | 1093 | |||
| GenBank mRNA | AK050135.1 | - | 5 | 396 | ||
| AK151863.1 | - | 11 | 1138 | |||
| AK160264.1 | - | 11 | 1090 | |||
| AY540996.1 | - | 11 | 1035 | |||
| AY540997.1 | - | 10 | 980 | |||
| BC009659.1 | - | 11 | 1058 | 3485959 | ||