Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102455.1 | NP_001095925.1 | 3, 4 | 491 | ||
NM_001102456.1 | NP_001095926.1 | 3, 4 | 491 | |||
NM_009691.2 | NP_033821.1 | 3, 4 | 491 | |||
GenBank mRNA | AK013376.1 | - | 3, 4 | 488 | ||
AK149977.1 | - | 3, 4 | 492 | |||
AK150020.1 | - | 3, 4 | 492 | |||
AK160471.1 | - | 3, 4 | 491 | |||
AK164012.1 | - | 3, 4 | 509 | |||
AK190139.1 | - | 3, 4 | 328 | |||
AK192625.1 | - | 3, 4 | 326 | |||
AK192927.1 | - | 3, 4 | 326 | |||
AK196095.1 | - | 2, 3 | 317 | |||
BC051999.1 | - | 3, 4 | 469 | 5716148 | ||
BC052396.1 | - | 3, 4 | 463 | 5719918 | ||
U15571.1 | - | 3, 4 | 467 | |||
Z22592.1 | - | 3, 4 | 491 | |||
Z27070.1 | - | 3, 4 | 489 |