Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111021.2 | NP_001104491.1 | 7 | 1026 | ||
NM_001111022.2 | NP_001104492.1 | 6 | 834 | |||
NM_001111023.2 | NP_001104493.1 | 5 | 2361 | |||
NM_009821.3 | NP_033951.2 | 4 | 2169 | |||
GenBank mRNA | AK051758.1 | - | 4 | 1707 | ||
AK137493.1 | - | 7 | 996 | |||
AK145098.1 | - | 7 | 1019 | |||
AK165307.1 | - | 7 | 1017 | |||
BC069929.1 | - | 6 | 771 | 4008335 | ||
D13802.1 | - | 5 | 1039 | |||
D26532.1 | - | 4 | 2166 |