Search
Search

Cck| Gene Symbol: Cck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284508.1 | NP_001271437.1 | 2 | 127 | ||
| NM_031161.4 | NP_112438.1 | 2 | 127 | |||
| GenBank mRNA | AK002677.1 | - | 2 | 95 | ||
| BC028487.1 | - | 2 | 72 | 1400830 | ||
| BY127153.1 | - | 2 | 96 | |||
| BY262295.1 | - | 2 | 123 | |||