Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020260.2 | NP_064656.2 | 6 | 976 | ||
GenBank mRNA | AF151363.1 | - | 6 | 617 | ||
AK018057.1 | - | 6 | 842 | |||
AK138417.1 | - | 6 | 1216 | |||
AK149975.1 | - | 6 | 812 | |||
AK150109.1 | - | 6 | 812 | |||
AK155729.1 | - | 6 | 796 | |||
AK170685.1 | - | 6 | 796 | |||
BC138870.1 | - | 6 | 781 | 8861892 |