Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271496.1 | NP_001258425.1 | 4 | 838 | ||
NM_013490.4 | NP_038518.2 | 3 | 784 | |||
GenBank mRNA | AB011002.1 | - | 3 | 643 | ||
AK005057.1 | - | 5 | 899 | |||
AK178697.1 | - | 2 | 54 | |||
AK182174.1 | - | 2 | 58 | |||
AK183570.1 | - | 2 | 58 | |||
AK186560.1 | - | 2 | 58 | |||
AK192596.1 | - | 2 | 54 | |||
AK215104.1 | - | 3 | 177 | |||
BC060218.1 | - | 3 | 747 |