siRNA Details
Gene Symbol: Elovl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007703.2 | NP_031729.1 | 4 | 634 | ||
GenBank mRNA | BC016468.1 | - | 4 | 629 | 4196583 | |
U97107.1 | - | 4 | 634 |

Gene Symbol: Elovl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007703.2 | NP_031729.1 | 4 | 634 | ||
GenBank mRNA | BC016468.1 | - | 4 | 629 | 4196583 | |
U97107.1 | - | 4 | 634 |