Search
Search

Dyrk1a| Gene Symbol: Dyrk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113389.1 | NP_001106860.1 | 10 | 1483 | ||
| NM_007890.2 | NP_031916.1 | 10 | 1742 | |||
| GenBank mRNA | AK052390.1 | - | 4 | 618 | ||
| BC034550.1 | - | 5 | 828 | |||
| BC048837.1 | - | 1 | 37 | |||
| BC129888.1 | - | 9 | 1345 | 40111469 | ||
| BC129889.1 | - | 9 | 1345 | 40111470 | ||
| BC138716.1 | - | 9 | 1372 | 8861738 | ||
| BC145405.1 | - | 9 | 1345 | 9053963 | ||
| U58497.1 | - | 9 | 1372 | |||