Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EEF2 SIRNA Assays ›
          Gene Symbol:
          Eef2 Ogdh
          Gene Name:
          eukaryotic translation elongation factor 2
          oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide)
          Gene Aliases:
          Ef-2
          2210403E04Rik, 2210412K19Rik, AA409584, d1401, mKIAA4192
          Chromosome Location:
          Chr.10: 81176631 - 81182509 on Build GRCm38
          Chromosome Location:
          Chr.11: 6291593 - 6359094 on Build GRCm38
          UniGene ID:
          Mm.2903
          Mm.276348
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID MSS274077
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Eef2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007907.2 NP_031933.1 5 854
          GenBank mRNA AK028938.1 - 5 833
          AK032997.1 - 5 840
          AK040474.1 - 5 840
          AK077866.1 - 5 840
          AK087985.1 - 5 840
          AK128976.1 - 5 839
          AK134195.1 - 5 840
          AK144874.1 - 5 842
          AK145509.1 - 5 840
          AK145545.1 - 5 840
          AK146508.1 - 5 840
          AK146947.1 - 5 840
          AK149659.1 - 5 840
          AK150136.1 - 5 840
          AK150902.1 - 5 840
          AK151487.1 - 5 840
          AK151812.1 - 5 790
          AK151945.1 - 5 840
          AK152146.1 - 5 840
          AK152447.1 - 5 840
          AK152587.1 - 5 840
          AK152826.1 - 5 840
          AK153275.1 - 5 840
          AK153936.1 - 5 840
          AK155325.1 - 5 840
          AK159155.1 - 5 841
          AK159421.1 - 5 840
          AK159514.1 - 5 840
          AK159806.1 - 5 840
          AK159829.1 - 5 840
          AK163063.1 - 5 840
          AK166212.1 - 5 840
          AK166595.1 - 5 840
          AK166596.1 - 5 840
          AK166719.1 - 5 840
          AK166747.1 - 5 840
          AK166751.1 - 5 838
          AK166758.1 - 5 840
          AK166788.1 - 5 840
          AK166815.1 - 5 840
          AK166829.1 - 5 840
          AK166851.1 - 5 838
          AK166968.1 - 5 840
          AK167081.1 - 5 840
          AK167093.1 - 5 840
          AK167109.1 - 5 840
          AK167115.1 - 5 840
          AK167199.1 - 5 840
          AK167208.1 - 5 838
          AK167221.1 - 5 840
          AK167230.1 - 5 840
          AK167281.1 - 5 839
          AK168527.1 - 5 840
          AK168615.1 - 5 840
          AK168638.1 - 5 840
          AK168659.1 - 5 840
          AK168794.1 - 5 840
          AK168827.1 - 5 840
          AK168874.1 - 5 833
          AK169013.1 - 5 840
          AK179811.1 - 1 73
          AK183998.1 - 1 73
          AK185420.1 - 2 207
          AK186796.1 - 1 73
          AK188086.1 - 1 73
          AK191500.1 - 2 201
          AK195289.1 - 1 73
          AK195475.1 - 1 73
          AK198227.1 - 1 73
          AK199770.1 - 1 72
          AK199882.1 - 1 73
          AK201728.1 - 1 73
          AK203781.1 - 374
          AK214102.1 - 1 72
          BC007152.1 - 5 838 3600352
          BC013263.1 - 5 826
          BC060707.1 - 4 715
          Gene Symbol: Ogdh
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK189909.1 - 30

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001961, X51466, Z11692, BC006547, BC136313, and others.

          Panther Classification:

          Molecular Function -

          translation elongation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          translation
          translational elongation
          positive regulation of translation
          nucleotide binding
          translation elongation factor activity
          GTPase activity
          protein binding
          GTP binding
          protein kinase binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0