Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EREG SIRNA Assays ›
Gene Symbol:
Ereg
Gene Name:
epiregulin
Gene Aliases:
EPR
Chromosome Location:
Chr.5: 91074617 - 91093649 on Build GRCm38
UniGene ID:
Mm.4791
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
MSS274180
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Contact Us
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ereg
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_007950.2
NP_031976.1
4
485
GenBank mRNA
BB612312.1
-
4
480
BC027838.1
-
4
461
5325124
D30782.1
-
4
480
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
angiogenesis
ovarian cumulus expansion
oocyte maturation
positive regulation of cytokine production
female meiotic division
epidermal growth factor receptor signaling pathway
cell-cell signaling
multicellular organism development
positive regulation of cell proliferation
negative regulation of cell proliferation
mRNA transcription
anatomical structure morphogenesis
organ morphogenesis
cytokine-mediated signaling pathway
cell differentiation
ovulation
positive regulation of cytokine biosynthetic process
positive regulation of phosphorylation
luteinizing hormone signaling pathway
keratinocyte proliferation
positive regulation of innate immune response
positive regulation of interleukin-6 biosynthetic process
positive regulation of DNA replication
positive regulation of epidermal growth factor-activated receptor activity
positive regulation of mitotic nuclear division
positive regulation of protein kinase activity
negative regulation of transcription, DNA-templated
positive regulation of fibroblast proliferation
primary follicle stage
positive regulation of smooth muscle cell proliferation
negative regulation of smooth muscle cell differentiation
positive regulation of cell division
epidermal growth factor receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-5x6th:80/100.66.78.110:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0