Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGF9 SIRNA Assays ›
          Gene Symbol:
          Fgf9
          Gene Name:
          fibroblast growth factor 9
          Gene Aliases:
          Eks
          Chromosome Location:
          Chr.14: 58071037 - 58112720 on Build GRCm38
          UniGene ID:
          Mm.8846
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS274340
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price Contact Us
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fgf9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013518.4 NP_038546.2 1 526
          GenBank mRNA AK158782.1 - 1 527
          AK164411.1 - 1 989
          BC099872.1 - 1 277 40040091
          BC099873.1 - 1 277 40040090
          BC099874.1 - 1 277 40040089
          BC099875.1 - 1 277 40040088
          BC125237.1 - 1 277 40040117
          D38258.1 - 1 238
          HM988990.1 - 1 156
          S82023.1 - 1 211
          U33535.1 - 1 294

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_002010, BC103979, D14838, BC069692, AK290792, and others.
          This siRNA also targets the following transcripts in Rat: NM_012952, D14839, BC078699.

          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          osteoblast differentiation
          positive regulation of mesenchymal cell proliferation
          chondrocyte differentiation
          cardiac left ventricle morphogenesis
          cardiac ventricle development
          protein import into nucleus
          cell-cell signaling
          multicellular organism development
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          male gonad development
          positive regulation of gene expression
          cell differentiation
          negative regulation of Wnt signaling pathway
          male sex determination
          lung development
          embryonic limb morphogenesis
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          positive regulation of activin receptor signaling pathway
          inner ear morphogenesis
          positive regulation of MAPK cascade
          positive regulation of smoothened signaling pathway
          positive regulation of transcription, DNA-templated
          regulation of timing of cell differentiation
          embryonic digestive tract development
          embryonic skeletal system development
          positive regulation of epithelial cell proliferation
          positive regulation of cell division
          positive regulation of cardiac muscle cell proliferation
          lung-associated mesenchyme development
          vasculogenesis involved in coronary vascular morphogenesis
          positive regulation of canonical Wnt signaling pathway
          receptor binding
          fibroblast growth factor receptor binding
          growth factor activity
          heparin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline