Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008141.3 | NP_032167.1 | 4 | 291 | ||
GenBank mRNA | AK009388.1 | - | 3 | 175 | ||
BC016272.1 | - | 4 | 289 | 4506728 | ||
BC085102.1 | - | 3 | 269 | |||
BC131941.1 | - | 4 | 286 | 40130218 | ||
BC131943.1 | - | 4 | 286 | 40130220 | ||
L10666.1 | - | 3 | 213 |